Product Details

SNP ID
rs6786491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:191202580 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCTAGCTTAATGTAATTATGATG[C/T]AAGATTTTGATCAAATATTTTATAT
Phenotype
MIM: 610280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OSTN PubMed Links

Gene Details

Gene
OSTN
Gene Name
osteocrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005247428.4 Intron XP_005247485.1
XM_017006303.1 Intron XP_016861792.1

View Full Product Details