Product Details

SNP ID
rs7920210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97684122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAGTGGGCCCTGGGGTTAGCAT[C/T]TGGTAGAAAACAGAACTCACAGCTC
Phenotype
MIM: 609763
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AVPI1 PubMed Links

Gene Details

Gene
AVPI1
Gene Name
arginine vasopressin induced 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021732.2 Intron NP_068378.2
XM_017016494.1 Intron XP_016871983.1
Gene
PI4K2A
Gene Name
phosphatidylinositol 4-kinase type 2 alpha
There are no transcripts associated with this gene.

View Full Product Details