Product Details

SNP ID
rs8121603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36579138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCCTGTCTCAAGGGAAGACAA[C/T]GAAATAGGGTCGCTGCAGTCAATTT
Phenotype
MIM: 607294
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DLGAP4-AS1 PubMed Links

Gene Details

Gene
DLGAP4-AS1
Gene Name
DLGAP4 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TGIF2
Gene Name
TGFB induced factor homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199513.1 Intron NP_001186442.1
NM_001199514.1 Intron NP_001186443.1
NM_001199515.1 Intron NP_001186444.1
NM_021809.6 Intron NP_068581.1
Gene
TGIF2-C20orf24
Gene Name
TGIF2-C20orf24 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199535.1 Intron NP_001186464.1

View Full Product Details