Product Details

SNP ID
rs4143230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:37258225 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGTGACCTCAGCGGGAAGGTGGC[C/G]GAAGATTGGGCGTAAAGGGGAGTGG
Phenotype
MIM: 616880
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TBC1D22B PubMed Links

Gene Details

Gene
TBC1D22B
Gene Name
TBC1 domain family member 22B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017772.3 178 Intron NP_060242.2
XM_011514738.2 178 Intron XP_011513040.1
XM_011514739.1 178 Intron XP_011513041.1
Gene
TMEM217
Gene Name
transmembrane protein 217
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162900.1 178 Intron NP_001156372.1
NM_001286401.1 178 Intron NP_001273330.1
NM_145316.3 178 Intron NP_660359.2
XM_011514362.1 178 Intron XP_011512664.1
XM_011514363.2 178 Intron XP_011512665.1
XM_011514364.2 178 UTR 5 XP_011512666.1
XM_011514365.2 178 Intron XP_011512667.1
XM_011514366.2 178 Intron XP_011512668.1
XM_011514367.2 178 Intron XP_011512669.1
XM_011514369.2 178 Intron XP_011512671.1
XM_011514370.2 178 Intron XP_011512672.1

View Full Product Details