Product Details

SNP ID
rs7184285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2840276 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACACATGTTCGAGTCCAGGAGGG[A/G]CACAGCCACCTCCTGAAGGACACTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRSS30P PubMed Links

Gene Details

Gene
PRSS30P
Gene Name
protease, serine, 30 pseudogene
There are no transcripts associated with this gene.

Gene
ZG16B
Gene Name
zymogen granule protein 16B
There are no transcripts associated with this gene.

View Full Product Details