Product Details

SNP ID
rs7838897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11349817 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATTGTATTTGTAGACCCTATTTT[A/C]TTTACCCGTTCATCTATTGGAGGCT
Phenotype
MIM: 615174
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TDH PubMed Links

Gene Details

Gene
TDH
Gene Name
L-threonine dehydrogenase (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details