Product Details
- SNP ID
-
rs6761110
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:88700721 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTAATGCAAGTACATCATAAAATA[C/G]TAGGAGCTGATGGAGGAAAGCAGTG
- Phenotype
-
MIM: 180430
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RPIA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75525908] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RPIA
- Gene Name
- ribose 5-phosphate isomerase A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144563.2 |
|
Intron |
|
|
NP_653164.2 |
View Full Product Details