Product Details

SNP ID
rs6587615
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:151726997 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTTGAGATCTGGTTCAGTGAAAA[C/T]GATCGTTGAGGCCGGGTGCCGTGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RIIAD1 PubMed Links

Gene Details

Gene
RIIAD1
Gene Name
regulatory subunit of type II PKA R-subunit (RIIa) domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144956.1 Intron NP_001138428.1

View Full Product Details