Product Details

SNP ID
rs7254161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36384808 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGTCTTCTTCATGTTACATTTTAC[C/T]GTTTACAAATTAAAAACCATGTGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFP14 PubMed Links

Gene Details

Gene
ZFP14
Gene Name
ZFP14 zinc finger protein
There are no transcripts associated with this gene.

Gene
ZFP82
Gene Name
ZFP82 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321917.1 Intron NP_001308846.1
NM_001321918.1 Intron NP_001308847.1
NM_001321919.1 Intron NP_001308848.1
NM_133466.3 Intron NP_597723.1

View Full Product Details