Product Details

SNP ID
rs6512309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:64261231 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTTTTAACTCAGTAGGGTGGAAA[A/G]GTTTTGCCGGAGGGGTGATAGGAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCMTD2 PubMed Links

Gene Details

Gene
PCMTD2
Gene Name
protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104925.1 Intron NP_001098395.1
NM_018257.2 Intron NP_060727.2

View Full Product Details