Product Details

SNP ID
rs7100497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133421232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGCCCCTGAGTGCCACCTGCATC[A/G]TGCCATAACCCTGACCGCCTGGGGC
Phenotype
MIM: 610447
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTG1 PubMed Links

Gene Details

Gene
MTG1
Gene Name
mitochondrial ribosome associated GTPase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138384.2 2607 Intron NP_612393.2
Gene
SPRN
Gene Name
shadow of prion protein homolog (zebrafish)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012508.3 2607 UTR 3 NP_001012526.2

View Full Product Details