Product Details
- SNP ID
-
rs7811392
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:128303810 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGGTCTCCATAGGTCATTATTCTT[A/C]CCAGGCGGGGTCAACTTCCTCTATT
- Phenotype
-
MIM: 612074
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MGC27345
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113169335] are located under a probe and SNP(s) [rs78629780] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MGC27345
- Gene Name
- uncharacterized protein MGC27345
There are no transcripts associated with this gene.
- Gene
- RBM28
- Gene Name
- RNA binding motif protein 28
View Full Product Details