Product Details

SNP ID
rs7953558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10246509 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCTTCACAAGATAGGGTAGGAAA[C/T]ATGATTTGTGACCTGTCTGAGAAAA
Phenotype
MIM: 602894
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLRD1 PubMed Links

Gene Details

Gene
KLRD1
Gene Name
killer cell lectin like receptor D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114396.1 Intron NP_001107868.1
NM_002262.3 Intron NP_002253.1
NM_007334.2 Intron NP_031360.1
XM_006719067.3 Intron XP_006719130.1
XM_011520650.2 Intron XP_011518952.1
XM_011520651.2 Intron XP_011518953.1
XM_017019285.1 Intron XP_016874774.1
XM_017019286.1 Intron XP_016874775.1
XM_017019287.1 Intron XP_016874776.1
XM_017019288.1 Intron XP_016874777.1
XM_017019289.1 Intron XP_016874778.1
XM_017019290.1 Intron XP_016874779.1
XM_017019291.1 Intron XP_016874780.1

View Full Product Details