Product Details
- SNP ID
-
rs7129669
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:121105216 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCAAGGACCCAACCCTCTGACCCC[A/G]GAGGATCTCATGTCTGTCTTTTGTA
- Phenotype
-
MIM: 602574
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TECTA
PubMed Links
Gene Details
- Gene
- TECTA
- Gene Name
- tectorin alpha
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005422.2 |
|
Intron |
|
|
NP_005413.2 |
View Full Product Details