Product Details
- SNP ID
-
rs4748021
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:6346106 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGCACAAACATAAAAACAGATCTT[A/G]TAGAAATATATATTTGATGGGCTTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LOC399716
PubMed Links
Gene Details
- Gene
- LOC399716
- Gene Name
- uncharacterized LOC399716
There are no transcripts associated with this gene.
View Full Product Details