Product Details
- SNP ID
-
rs7023344
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:122821959 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTACTTTATCATAACCAGCAACATT[C/T]TGAGTTCACTATTTTACTTATTGCC
- Phenotype
-
MIM: 604421
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PDCL
PubMed Links
Gene Details
- Gene
- PDCL
- Gene Name
- phosducin like
View Full Product Details