Product Details

SNP ID
rs7283462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:32249069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAACCTAAGAGCACTTGCATAG[A/C]TGTCATTAAACTTCCCAGGCTGATC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIS18A PubMed Links

Gene Details

Gene
MIS18A
Gene Name
MIS18 kinetochore protein A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018944.2 Intron NP_061817.1
XM_017028400.1 Intron XP_016883889.1
XM_017028401.1 Intron XP_016883890.1
XM_017028402.1 Intron XP_016883891.1

View Full Product Details