Product Details

SNP ID
rs10191914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:102314197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAAGACTGCTATGTGCCAGACCC[C/T]GAGACAAGCACTTTCCACACACCAC
Phenotype
MIM: 601203
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL1RL1 PubMed Links

Gene Details

Gene
IL1RL1
Gene Name
interleukin 1 receptor like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282408.1 Intron NP_001269337.1
NM_003856.3 Intron NP_003847.2
NM_016232.4 Intron NP_057316.3
XM_006712839.3 Intron XP_006712902.1
XM_011512151.1 Intron XP_011510453.1

View Full Product Details