Product Details

SNP ID
rs10416166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49977897 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCACCCCTCACCACTGGCCCACTC[C/T]GTGCGCAGGTGCTCCACGAATCGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIGLEC16 PubMed Links

Gene Details

Gene
SIGLEC16
Gene Name
sialic acid binding Ig like lectin 16 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
VRK3
Gene Name
vaccinia related kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025778.1 Intron NP_001020949.1
NM_001308420.1 Intron NP_001295349.1
NM_016440.3 Intron NP_057524.3
XM_005258971.3 Intron XP_005259028.1
XM_005258972.3 Intron XP_005259029.1
XM_006723237.3 Intron XP_006723300.1
XM_006723238.3 Intron XP_006723301.1
XM_011527023.2 Intron XP_011525325.1

View Full Product Details