Product Details

SNP ID
rs3118766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:94263016 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAGTTATTTCACAACCCAGCATG[G/T]AGTGTGAGTTGGGGAATGTACCATG
Phenotype
MIM: 603404
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF169 PubMed Links

Gene Details

Gene
ZNF169
Gene Name
zinc finger protein 169
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301275.1 Intron NP_001288204.1
NM_003448.2 Intron NP_003439.2
NM_194320.3 Intron NP_919301.2
XM_017014362.1 Intron XP_016869851.1
XM_017014363.1 Intron XP_016869852.1
XM_017014364.1 Intron XP_016869853.1
XM_017014365.1 Intron XP_016869854.1
XM_017014366.1 Intron XP_016869855.1

View Full Product Details