Product Details

SNP ID
rs9976062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:44573798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCGACCTGAGCTACAGCAGCCGC[A/G]TCTGCCTTCCTGGTTCCTGTGACTC
Phenotype
MIM: 612920
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP10-4 PubMed Links

Gene Details

Gene
KRTAP10-4
Gene Name
keratin associated protein 10-4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198687.2 70 Missense Mutation ATC,GTC I14V NP_941960.2
Gene
KRTAP10-5
Gene Name
keratin associated protein 10-5
There are no transcripts associated with this gene.

Gene
TSPEAR
Gene Name
thrombospondin type laminin G domain and EAR repeats
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272037.1 70 Intron NP_001258966.1
NM_144991.2 70 Intron NP_659428.2

View Full Product Details