Product Details

SNP ID
rs9939401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75152163 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTTGGTTTTCATTTGTTTGGATA[C/T]GATGTGTCCATGTGTGTTTTTGTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFP1 PubMed Links

Gene Details

Gene
ZFP1
Gene Name
ZFP1 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318469.1 Intron NP_001305398.1
NM_001318471.1 Intron NP_001305400.1
NM_001318472.1 Intron NP_001305401.1
NM_001318473.1 Intron NP_001305402.1
NM_001318474.1 Intron NP_001305403.1
NM_001318475.1 Intron NP_001305404.1
NM_001318476.1 Intron NP_001305405.1
NM_153688.3 Intron NP_710155.2
XM_011522921.2 Intron XP_011521223.1
XM_017022983.1 Intron XP_016878472.1
XM_017022984.1 Intron XP_016878473.1
XM_017022985.1 Intron XP_016878474.1
XM_017022986.1 Intron XP_016878475.1
XM_017022987.1 Intron XP_016878476.1

View Full Product Details