Product Details

SNP ID
rs9305332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:27735685 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAATTGAACAGAGACCACTGACC[A/G]TAAGAGACAATATGAATTGGAGAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00113 PubMed Links

Gene Details

Gene
LINC00113
Gene Name
long intergenic non-protein coding RNA 113
There are no transcripts associated with this gene.

View Full Product Details