Product Details

SNP ID
rs9891706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:60690032 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTAGGCAGAAAAATGGTTTTCT[A/T]AAAAAAGGAGTTATTCAAGTTTGGT
Phenotype
MIM: 607470
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BCAS3 PubMed Links

Gene Details

Gene
BCAS3
Gene Name
BCAS3, microtubule associated cell migration factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099432.1 Intron NP_001092902.1
NM_001320470.1 Intron NP_001307399.1
NM_017679.3 Intron NP_060149.3
XM_005257472.1 Intron XP_005257529.1
XM_011524939.1 Intron XP_011523241.1
XM_011524940.1 Intron XP_011523242.1
XM_011524941.1 Intron XP_011523243.1
XM_011524942.2 Intron XP_011523244.1
XM_011524943.2 Intron XP_011523245.1
XM_011524944.1 Intron XP_011523246.1
XM_011524945.2 Intron XP_011523247.1
XM_011524946.2 Intron XP_011523248.1
XM_011524947.1 Intron XP_011523249.1
XM_011524948.2 Intron XP_011523250.1
XM_011524949.2 Intron XP_011523251.1
XM_011524950.1 Intron XP_011523252.1
XM_011524951.2 Intron XP_011523253.1
XM_017024783.1 Intron XP_016880272.1
XM_017024784.1 Intron XP_016880273.1
XM_017024785.1 Intron XP_016880274.1
XM_017024786.1 Intron XP_016880275.1
XM_017024787.1 Intron XP_016880276.1
XM_017024788.1 Intron XP_016880277.1
XM_017024789.1 Intron XP_016880278.1
XM_017024790.1 Intron XP_016880279.1
XM_017024791.1 Intron XP_016880280.1
XM_017024792.1 Intron XP_016880281.1
XM_017024793.1 Intron XP_016880282.1
XM_017024794.1 Intron XP_016880283.1
XM_017024795.1 Intron XP_016880284.1
XM_017024796.1 Intron XP_016880285.1

View Full Product Details