Product Details
- SNP ID
-
rs9622242
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:35656363 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATATGTGCAGTAACGTTGTTTTT[C/G]TACATTCCTGACCAGAAAATCATTT
- Phenotype
-
MIM: 607256
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
APOL6
PubMed Links
Gene Details
- Gene
- APOL6
- Gene Name
- apolipoprotein L6
View Full Product Details