Product Details
- SNP ID
-
rs5981075
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:71122583 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCAGTTGAAGTTCGCTGGTCTTTC[C/G]ATAAATGCCAGGAAGCTACTGCAGG
- Phenotype
-
MIM: 300188
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MED12
PubMed Links
Gene Details
- Gene
- MED12
- Gene Name
- mediator complex subunit 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005120.2 |
1523 |
Missense Mutation |
CAT,GAT |
H442D |
NP_005111.2 |
View Full Product Details