Product Details

SNP ID
rs6061469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62246779 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCACACCAGGTTCTCCCTGGGTCC[A/G]TTCATGTCACAGCAGGCTTTGTCTT
Phenotype
MIM: 606731
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OSBPL2 PubMed Links

Gene Details

Gene
OSBPL2
Gene Name
oxysterol binding protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278649.1 Intron NP_001265578.1
NM_014835.3 Intron NP_055650.1
NM_144498.2 Intron NP_653081.1
XM_017028163.1 Intron XP_016883652.1
XM_017028164.1 Intron XP_016883653.1
XM_017028165.1 Intron XP_016883654.1
XM_017028166.1 Intron XP_016883655.1
XM_017028167.1 Intron XP_016883656.1
XM_017028168.1 Intron XP_016883657.1
XM_017028169.1 Intron XP_016883658.1
XM_017028170.1 Intron XP_016883659.1

View Full Product Details