Product Details

SNP ID
rs10138696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23519268 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACCACACTTCTAATCATTCCATG[C/T]CAGGTGTTTGTCGATGCTTTCTTTA
Phenotype
MIM: 611612
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
THTPA PubMed Links

Gene Details

Gene
THTPA
Gene Name
thiamine triphosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126339.3 Intron NP_001119811.1
NM_001256062.2 Intron NP_001242991.1
NM_001256321.2 Intron NP_001243250.1
NM_001256322.2 Intron NP_001243251.1
NM_001256323.2 Intron NP_001243252.1
NM_024328.5 Intron NP_077304.1
Gene
ZFHX2
Gene Name
zinc finger homeobox 2
There are no transcripts associated with this gene.

View Full Product Details