Product Details

SNP ID
rs7526578
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:54030807 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGCTTTTCATTTGGACTCAGGAA[C/G]GCTAAAGACTAGGCCATAGGAGGAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM59 PubMed Links

Gene Details

Gene
TMEM59
Gene Name
transmembrane protein 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305043.1 2238 UTR 3 NP_001291972.1
NM_001305049.1 2238 UTR 3 NP_001291978.1
NM_001305050.1 2238 UTR 3 NP_001291979.1
NM_001305051.1 2238 UTR 3 NP_001291980.1
NM_001305052.1 2238 UTR 3 NP_001291981.1
NM_001305066.1 2238 Intron NP_001291995.1
NM_004872.4 2238 UTR 3 NP_004863.2
XM_006711051.1 2238 Intron XP_006711114.1

View Full Product Details