Product Details
- SNP ID
-
rs10427608
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:23359943 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTGAGTCTTAGAGAGGTGAAGTCG[C/T]TGGGCCCACCTGGAAGAACCATGAT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CES5AP1
PubMed Links
Gene Details
- Gene
- CES5AP1
- Gene Name
- carboxylesterase 5A pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details