Product Details

SNP ID
rs9930995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:22300268 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGTTATGCAACTGTATGTATGTT[C/G]TGGGTTATTTATATGAGATGGATTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
POLR3E PubMed Links

Gene Details

Gene
POLR3E
Gene Name
RNA polymerase III subunit E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258033.1 Intron NP_001244962.1
NM_001258034.1 Intron NP_001244963.1
NM_001258035.1 Intron NP_001244964.1
NM_001258036.1 Intron NP_001244965.1
NM_018119.3 Intron NP_060589.1
XM_006721063.2 Intron XP_006721126.1
XM_011545892.1 Intron XP_011544194.1
XM_011545893.2 Intron XP_011544195.1
XM_017023443.1 Intron XP_016878932.1

View Full Product Details