Product Details

SNP ID
rs9991837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68227061 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTCTCTGGTCTTCAGTTTCTTGA[A/T]CTGTAAATTGAAAGACTCAGTGTAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TMPRSS11B PubMed Links

Gene Details

Gene
TMPRSS11B
Gene Name
transmembrane protease, serine 11B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182502.3 2237 UTR 3 NP_872308.2
XM_011531608.2 2237 UTR 3 XP_011529910.1
XM_011531609.1 2237 Intron XP_011529911.1
Gene
TMPRSS11BNL
Gene Name
TMPRSS11B N-terminal like, pseudogene
There are no transcripts associated with this gene.

View Full Product Details