Product Details

SNP ID
rs9611112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:39223997 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATCAAAGATGGGGAAGTGTGCAA[A/G]CAGGGAGGGGTGCGGGCAAGAGGGC
Phenotype
MIM: 190040
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDGFB PubMed Links

Gene Details

Gene
PDGFB
Gene Name
platelet derived growth factor subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002608.3 3163 UTR 3 NP_002599.1
NM_033016.3 3163 UTR 3 NP_148937.1

View Full Product Details