Product Details

SNP ID
rs9753600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36894870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCATCTCTATTTCTACCCAGCTT[G/T]CTTTCTTTCTTTCTTTTTTTGAGAC
Phenotype
MIM: 606754
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SAMHD1 PubMed Links

Gene Details

Gene
SAMHD1
Gene Name
SAM and HD domain containing deoxynucleoside triphosphate triphosphohydrolase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015474.3 Intron NP_056289.2
XM_005260384.3 Intron XP_005260441.1
XM_011528761.2 Intron XP_011527063.1
Gene
TLDC2
Gene Name
TBC/LysM-associated domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304783.1 Intron NP_001291712.1
NM_080628.2 Intron NP_542195.1
XM_017027672.1 Intron XP_016883161.1
XM_017027673.1 Intron XP_016883162.1
XM_017027674.1 Intron XP_016883163.1

View Full Product Details