Product Details

SNP ID
rs9823824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112463268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGTACGGCACTCTGGAAAAGGC[A/G]AAGCAATAGAGGCAGTGGTTGTCAG
Phenotype
MIM: 607925
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTLA PubMed Links

Gene Details

Gene
BTLA
Gene Name
B and T lymphocyte associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085357.1 4025 Intron NP_001078826.1
NM_181780.3 4025 Intron NP_861445.3
XM_011512447.2 4025 UTR 3 XP_011510749.1
XM_017005748.1 4025 UTR 3 XP_016861237.1

View Full Product Details