Product Details
- SNP ID
-
rs10411092
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:16482721 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCATCGGCGCTGGCCAGTCCCCA[C/T]CCAGGTCACCGTTCCAGTCGCTCTG
- Phenotype
-
MIM: 611414
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CALR3
PubMed Links
Gene Details
- Gene
- CALR3
- Gene Name
- calreticulin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145046.4 |
806 |
Missense Mutation |
|
|
NP_659483.2 |
View Full Product Details