Product Details

SNP ID
rs6002485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:22486707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGGGGCCCTTCACCAAACAAAA[C/G]CATTTTACTTTGAATGGTTTGGCTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF280B PubMed Links

Gene Details

Gene
ZNF280B
Gene Name
zinc finger protein 280B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080764.3 3101 UTR 3 NP_542942.2
XM_011529893.2 3101 UTR 3 XP_011528195.1
XM_011529895.2 3101 UTR 3 XP_011528197.1
XM_011529896.2 3101 UTR 3 XP_011528198.1
XM_011529897.2 3101 Intron XP_011528199.1

View Full Product Details