Product Details

SNP ID
rs5951686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:22034192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAAACTCTTGTAAGATTTCTCA[A/G]CACTTAGGGTTTAAACAAGGGTCCT
Phenotype
MIM: 300550
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHEX PubMed Links

Gene Details

Gene
PHEX
Gene Name
phosphate regulating endopeptidase homolog, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000444.5 Intron NP_000435.3
NM_001282754.1 Intron NP_001269683.1
XM_011545533.1 Intron XP_011543835.1
XM_011545536.2 Intron XP_011543838.1
XM_017029579.1 Intron XP_016885068.1

View Full Product Details