Product Details

SNP ID
rs7526192
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:38889993 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTCAATATTCATTTAGTTGTAAA[C/T]GACTATTACACAAAGTGCTAGATTC
Phenotype
MIM: 611923 MIM: 608962
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GJA9 PubMed Links

Gene Details

Gene
GJA9
Gene Name
gap junction protein alpha 9
There are no transcripts associated with this gene.

Gene
GJA9-MYCBP
Gene Name
GJA9-MYCBP readthrough
There are no transcripts associated with this gene.

Gene
RHBDL2
Gene Name
rhomboid like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304746.1 Intron NP_001291675.1
NM_017821.4 Intron NP_060291.2

View Full Product Details