Product Details
- SNP ID
-
rs4890128
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40749901 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTAACATCAAGAATGGATAAAATA[A/G]TAACTGCCAAGGGAAGTGGGGCACA
- Phenotype
-
MIM: 616646
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT25
PubMed Links
Gene Details
- Gene
- KRT25
- Gene Name
- keratin 25
View Full Product Details