Product Details

SNP ID
rs9462981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:44346770 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTTTCATCTTATAAATTTATAAA[A/T]GCTCAAGGAGTGTAAATTAGTTCAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SPATS1 PubMed Links

Gene Details

Gene
SPATS1
Gene Name
spermatogenesis associated serine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145026.3 Intron NP_659463.1

View Full Product Details