Product Details

SNP ID
rs9645801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56320060 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAGAGGACACAGGGCTTGGATAG[A/G]GAAGTGACTAGGGGAGAGAAGCCCA
Phenotype
MIM: 605861
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNPY2 PubMed Links

Gene Details

Gene
CNPY2
Gene Name
canopy FGF signaling regulator 2
There are no transcripts associated with this gene.

Gene
PAN2
Gene Name
PAN2 poly(A) specific ribonuclease subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127460.2 Intron NP_001120932.1
NM_001166279.1 Intron NP_001159751.1
NM_014871.4 Intron NP_055686.3

View Full Product Details