Product Details

SNP ID
rs9886692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35419270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGTATTCCACTGACAGTGCTAG[A/G]GAGATCATCAAGACAAAGTCAACAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP8B5P PubMed Links

Gene Details

Gene
ATP8B5P
Gene Name
ATPase phospholipid transporting 8B5, pseudogene
There are no transcripts associated with this gene.

View Full Product Details