Product Details

SNP ID
rs6102586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36878609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAAAAAGGAAAGAAACTAATTC[C/T]TACCTAGAACCAGCCCTTGGGAACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TLDC2 PubMed Links

Gene Details

Gene
TLDC2
Gene Name
TBC/LysM-associated domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304783.1 Intron NP_001291712.1
NM_080628.2 Intron NP_542195.1

View Full Product Details