Product Details

SNP ID
rs10223397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:85611996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAACATCTGATGTCTTACAAAA[A/G]TAACATCTTGCTAAACTATTATACA
Phenotype
MIM: 616686
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SYNCRIP PubMed Links

Gene Details

Gene
SYNCRIP
Gene Name
synaptotagmin binding cytoplasmic RNA interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159673.1 5024 UTR 3 NP_001153145.1
NM_001159674.1 5024 UTR 3 NP_001153146.1
NM_001159675.1 5024 Intron NP_001153147.1
NM_001159676.1 5024 UTR 3 NP_001153148.1
NM_001159677.1 5024 UTR 3 NP_001153149.1
NM_001253771.1 5024 UTR 3 NP_001240700.1
NM_006372.4 5024 Intron NP_006363.4
XM_005248635.4 5024 UTR 3 XP_005248692.1
XM_005248636.2 5024 UTR 3 XP_005248693.1
XM_005248637.2 5024 UTR 3 XP_005248694.1
XM_017010176.1 5024 UTR 3 XP_016865665.1
XM_017010177.1 5024 UTR 3 XP_016865666.1
XM_017010178.1 5024 UTR 3 XP_016865667.1
XM_017010179.1 5024 UTR 3 XP_016865668.1
XM_017010180.1 5024 Intron XP_016865669.1

View Full Product Details