Product Details
- SNP ID
-
rs9375666
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:129831307 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTATTCTATTTAACATTATTTCTGT[A/G]CATTAGAGAATCTAAACAAGCAATT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM244
PubMed Links
Gene Details
- Gene
- TMEM244
- Gene Name
- transmembrane protein 244
View Full Product Details