Product Details

SNP ID
rs9832625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:29287529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAGATATTTGGAAATATGATGCT[A/C]TTCAGTGTTGTATAAATATTGCTCT
Phenotype
MIM: 605786
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RBMS3 PubMed Links

Gene Details

Gene
RBMS3
Gene Name
RNA binding motif single stranded interacting protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003792.2 Intron NP_001003792.1
NM_001003793.2 Intron NP_001003793.1
NM_001177711.1 Intron NP_001171182.1
NM_001177712.1 Intron NP_001171183.1
NM_014483.3 Intron NP_055298.2
XM_005265060.2 Intron XP_005265117.1
XM_005265061.2 Intron XP_005265118.1
XM_005265062.1 Intron XP_005265119.1
XM_005265063.1 Intron XP_005265120.1
XM_005265065.4 Intron XP_005265122.1
XM_017006178.1 Intron XP_016861667.1
XM_017006179.1 Intron XP_016861668.1
XM_017006180.1 Intron XP_016861669.1
XM_017006181.1 Intron XP_016861670.1
XM_017006182.1 Intron XP_016861671.1
Gene
RBMS3-AS3
Gene Name
RBMS3 antisense RNA 3
There are no transcripts associated with this gene.

View Full Product Details