Product Details

SNP ID
rs6705235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:61486445 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTGTATATGTAAGATGGCAGCAA[C/G]CGAGCCCTCAATACTGTCTTTTTAT
Phenotype
MIM: 602559
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
XPO1 PubMed Links

Gene Details

Gene
XPO1
Gene Name
exportin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003400.3 Intron NP_003391.1
XM_005264544.1 Intron XP_005264601.1
XM_005264546.1 Intron XP_005264603.1
XM_006712094.2 Intron XP_006712157.1
XM_011533097.1 Intron XP_011531399.1
XM_011533098.1 Intron XP_011531400.1
XM_011533099.2 Intron XP_011531401.1

View Full Product Details