Product Details

SNP ID
rs6750795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231513520 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTACTCTGCTCAACTCTGTACATA[C/T]AATTACCTCATCTCTACTGCCATTG
Phenotype
MIM: 604153
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00471 PubMed Links

Gene Details

Gene
LINC00471
Gene Name
long intergenic non-protein coding RNA 471
There are no transcripts associated with this gene.

Gene
NMUR1
Gene Name
neuromedin U receptor 1
There are no transcripts associated with this gene.

View Full Product Details