Product Details

SNP ID
rs9458079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:161006585 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTTTCCAAGGGAGGTCCTAGAAT[C/T]AATCCCTCGTAGATACTGAAGGATG
Phenotype
MIM: 602425
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP3K4 PubMed Links

Gene Details

Gene
MAP3K4
Gene Name
mitogen-activated protein kinase kinase kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291958.1 Intron NP_001278887.1
NM_001301072.1 Intron NP_001288001.1
NM_005922.3 Intron NP_005913.2
NM_006724.3 Intron NP_006715.2
XM_005266989.2 Intron XP_005267046.1
XM_017010869.1 Intron XP_016866358.1

View Full Product Details